missing translation for 'onlineSavingsMsg'
Learn More

1-Iodopentane, 97%, stabilized

Product Code. 13428558 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
13428558 500 g Glass bottle 500g
10321301 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13428558 Supplier Thermo Scientific Acros Supplier No. 412475000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 628-17-1 | C5H11I | 198.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-17-1
Molecular Formula C5H11I
Molecular Weight (g/mol) 198.05
MDL Number MFCD00001100
InChI Key BLXSFCHWMBESKV-UHFFFAOYSA-N
Synonym n-amyl iodide, amyl iodide, pentyl iodide, n-pentyl iodide, pentane, 1-iodo, 1-pentyl iodide, 1-jodpentan, 1-jodpentan czech, iodopentane, pentyliodide
PubChem CID 12335
IUPAC Name 1-iodopentane
SMILES CCCCCI

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.5100g/mL
Boiling Point 155.0°C to 157.0°C
Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)4I
Refractive Index 1.4940 to 1.4960
Quantity 500 g
Beilstein 01, 133
Specific Gravity 1.51
Solubility Information Solubility in water: insoluble
Stabilizer copper
Formula Weight 198.05
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Iodopentane, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Iodopentane, stabilized
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.