missing translation for 'onlineSavingsMsg'
Learn More

1-Iodonaphthalene, 97.5%

Product Code. 10360304 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Unit Size:
10mL
50mL
This item is not returnable. View return policy

Product Code. 10360304

Brand: Thermo Scientific Acros 218420500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 90-14-2 | C10H7I | 254.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-14-2
Molecular Formula C10H7I
Molecular Weight (g/mol) 254.07
MDL Number MFCD00003876
InChI Key NHPPIJMARIVBGU-UHFFFAOYSA-N
Synonym naphthalene, 1-iodo, 1-naphthyl iodide, alpha-iodonaphthalene, iodonaphthalene, 1-iodonaphthalenen, .alpha.-iodonaphthalene, 1-iodo-naphthalene, 1-iodonaphthlen, 1-iodonaphthlene, 1-iodonapthalene
PubChem CID 7004
IUPAC Name 1-iodonaphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2I

Specifications

CAS Min % 96.5
CAS Max % 100.0
Color Yellow to Brown
Density 1.7400g/mL
Boiling Point 302.0°C to 305.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96.5% min. (GC)
Packaging Glass bottle
Linear Formula C10H7I
Refractive Index 1.6980 to 1.7040
Quantity 50 mL
Beilstein 05, 550
Specific Gravity 1.74
Formula Weight 254.07
Percent Purity 97.5%
Physical Form Liquid
Chemical Name or Material 1-Iodonaphthalene
Show More Show Less
compliance-icons
Product Identifier
  • 1-Iodonaphthalene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.