missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-4-nitrobenzene, TRC

Artikelnummer. 30408250
Ansicht ändern
Click to view available options
Quantity:
50 g
Packungsgröße:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packungsgröße
30408250 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30408250 Lieferant Toronto Research Chemicals Lieferanten-Nr. TRCI71563350G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Iodo-4-nitrobenzene
CAS 636-98-6
Molecular Formula C6H4INO2
Quantity 50 g
InChI Formula InChI=1S/C6H4INO2/c7-5-1-3-6(4-2-5)8(9)10/h1-4H
SMILES [O-][N+](=O)c1ccc(I)cc1
IUPAC Name 1-iodo-4-nitrobenzene
Molecular Weight (g/mol) 249.01
Formula Weight 248.9287
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.