missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-4-n-pentylbenzene, 98%

Product Code. 11347119
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11347119 10 g 10g
11337119 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347119 Supplier Thermo Scientific Alfa Aesar Supplier No. H27356.09
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 85017-60-3 | C11H15I | 274.145 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85017-60-3
Molecular Formula C11H15I
Molecular Weight (g/mol) 274.145
MDL Number MFCD00079767
InChI Key MTRMOYSZRPLAOL-UHFFFAOYSA-N
Synonym 4-iodopentylbenzene, 1-iodo-4-n-pentylbenzene, 4-pentyliodobenzene, #, 1-iodanyl-4-pentyl-benzene
PubChem CID 145057
IUPAC Name 1-iodo-4-pentylbenzene
SMILES CCCCCC1=CC=C(C=C1)I

Specifications

Density 1.38
Boiling Point 88°C (1 mmHg)
Flash Point >110°C (230°F)
Quantity 10 g
Sensitivity Light sensitive
Formula Weight 274.14
Percent Purity 98%
Chemical Name or Material 1-Iodo-4-n-pentylbenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.