missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-3-(trimethylsiloxy)benzene, 98%, Thermo Scientific™

Product Code. 11395355
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11395355 1 g 1g
11305365 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11395355 Supplier Thermo Scientific Alfa Aesar Supplier No. L19588.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 732306-25-1
Molecular Formula C9H13IOSi
Molecular Weight (g/mol) 292.191
MDL Number MFCD03788743
InChI Key GJDJYHHPFQRAHK-UHFFFAOYSA-N
Synonym 1-iodo-3-trimethylsiloxy benzene, 3-iodophenoxytrimethylsilane
PubChem CID 53411742
IUPAC Name (3-iodophenoxy)-trimethylsilane
SMILES C[Si](C)(C)OC1=CC(=CC=C1)I

Specifications

Quantity 1 g
Formula Weight 292.19
Percent Purity 98%
Refractive Index 1.546
Sensitivity Light and moisture sensitive
Chemical Name or Material 1-Iodo-3-(trimethylsiloxy)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.