missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-3-(trifluoromethoxy)benzene, 97+%, Thermo Scientific™

Catalog Number 11421197
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
198206-33-6
C7H4F3IO
288.01
MFCD01090992
UQZXQSQWKJZHCD-UHFFFAOYSA-N
1-iodo-3-trifluoromethoxy benzene, 3-trifluoromethoxy iodobenzene, 3-iodo-1-trifluoromethoxy benzene, m-iodotrifluoromethoxybenzene, 1-iodo-3-trifluoromethoxy-benzene, benzene, 1-iodo-3-trifluoromethoxy, pubchem4438, acmc-1c5ss, 3-trifluoromethoxyiodobenzene, ksc494i7t
2777293
1-iodo-3-(trifluoromethoxy)benzene
FC(F)(F)OC1=CC=CC(I)=C1
This item is not returnable. View return policy
198206-33-6
C7H4F3IO
288.01
MFCD01090992
UQZXQSQWKJZHCD-UHFFFAOYSA-N
1-iodo-3-trifluoromethoxy benzene, 3-trifluoromethoxy iodobenzene, 3-iodo-1-trifluoromethoxy benzene, m-iodotrifluoromethoxybenzene, 1-iodo-3-trifluoromethoxy-benzene, benzene, 1-iodo-3-trifluoromethoxy, pubchem4438, acmc-1c5ss, 3-trifluoromethoxyiodobenzene, ksc494i7t
2777293
1-iodo-3-(trifluoromethoxy)benzene
FC(F)(F)OC1=CC=CC(I)=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 1.855
Boiling Point 185°C to 186°C
Flash Point 65°C (150°F)
Refractive Index 1.52
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 288.01
Percent Purity ≥97%
Chemical Name or Material 1-Iodo-3-(trifluoromethoxy)benzene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'