missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-3-nitrobenzene, 99%

Product Code. 11447224
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11447224 500 g 500g
11427224 25 g 25g
11437224 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11447224 Supplier Thermo Scientific Alfa Aesar Supplier No. B21605.36

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 645-00-1 | C6H4INO2 | 249.007 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 645-00-1
Molecular Formula C6H4INO2
Molecular Weight (g/mol) 249.007
MDL Number MFCD00007218
InChI Key CBYAZOKPJYBCHE-UHFFFAOYSA-N
Synonym 3-iodonitrobenzene, m-iodonitrobenzene, benzene, 1-iodo-3-nitro, 3-nitroiodobenzene, meta-iodonitrobenzene, 3-i-nitrobenzene, iodo-3-nitrobenzene, meta-nitroiodobenzene, 1-iodo-3-nitro-benzene, 3-nitro-iodobenzen
PubChem CID 12574
ChEBI CHEBI:67123
IUPAC Name 1-iodo-3-nitrobenzene
SMILES C1=CC(=CC(=C1)I)[N+](=O)[O-]

Specifications

Melting Point 35°C to 38°C
Boiling Point 279°C to 280°C
Flash Point 71°C (159°F)
Quantity 500 g
UN Number UN2926
Beilstein 1525167
Formula Weight 249.01
Percent Purity 99%
Chemical Name or Material 1-Iodo-3-nitrobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.