missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-2-(trimethylsilyl)acetylene, 97%

Product Code. p-7035656
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15460128

Brand: Thermo Scientific Alfa Aesar H53375.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18163-47-8 | C5H9ISi | 224.12 g/mol

1-Iodo-2-(trimethylsilyl)acetylene may be used to investigate the reactivity of diamagnetic cobalt(I) complex, Tol-BDI((2-pp)2)Co [Tol-BDI((2-pp)2)H = 2-(4-tolyl)-1,3-bis(2-isopropylpyridyl)propenediimine]. It may be used for the synthesis of 1-(4-trimethylsilanylbuta-1,3-diynyl)pyrene, via Pd-coupling with 1-iodo-2-trimethylsilylacetylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Iodo-2-(trimethylsilyl)acetylene may be used to investigate the reactivity of diamagnetic cobalt(I) complex, Tol-BDI((2-pp)2)Co [Tol-BDI((2-pp)2)H = 2-(4-tolyl)-1,3-bis(2-isopropylpyridyl)propenediimine]. It may be used for the synthesis of 1-(4-trimethylsilanylbuta-1,3-diynyl)pyrene, via Pd-coupling with 1-iodo-2-trimethylsilylacetylene.

Notes
Light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18163-47-8
Molecular Formula C5H9ISi
Molecular Weight (g/mol) 224.12
MDL Number MFCD00274201
InChI Key HNIRHTRSZDSMOF-UHFFFAOYSA-N
Synonym 1-iodo-2-trimethylsilyl acetylene, iodoethynyl trimethylsilane, silane, iodomethyl trimethyl, 2-iodoethynyl trimethylsilane, 2-iodoethynyl trimethyl silane, silane, iodoethynyl trimethyl, tbdmcs, acmc-20aoug, trimethyliodoethinylsilane, iodoethynyltrimethylsilane
PubChem CID 140341
IUPAC Name 2-iodoethynyl(trimethyl)silane
SMILES C[Si](C)(C)C#CI

Specifications

Boiling Point 130°C
Quantity 1 g
Linear Formula (CH3)3SiC≡CI
UN Number UN1993
Flash Point 48°C (118°F)
Formula Weight 224.12
Percent Purity 97%
Refractive Index 1.511
Sensitivity Light sensitive
Density 1.46
Chemical Name or Material 1-Iodo-2-(trimethylsilyl)acetylene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.