missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-2,4-dinitrobenzene, 98%

Product Code. 11480058
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480058 5 g 5g
11490058 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11480058 Supplier Thermo Scientific Alfa Aesar Supplier No. B23659.06
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 709-49-9 | C6H3IN2O4 | 294.004 g/mol

It is employed in the cloning and characterization of a biotic-stress-inducible glutathione transferase from Phaseolus vulgaris.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the cloning and characterization of a biotic-stress-inducible glutathione transferase from Phaseolus vulgaris.

Solubility
Insoluble in water.

Notes
Light sensitive. Store away from oxidizing agents, bases and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under dark.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 709-49-9
Molecular Formula C6H3IN2O4
Molecular Weight (g/mol) 294.004
MDL Number MFCD00039738
InChI Key FXMKXMJLXRTQSW-UHFFFAOYSA-N
Synonym 2,4-dinitroiodobenzene, benzene, 1-iodo-2,4-dinitro, unii-not697x2xm, ccris 7907, 4-iodo-1,3-dintrobenzene, 6-iodo-1,3-dintrobenzene, 2,4-dinitro-1-iodobenzene, not697x2xm, 1-iodo-2,4-dinitro-benzene, 1-iod-2,4-dinitrobenzol
PubChem CID 69730
ChEBI CHEBI:59000
IUPAC Name 1-iodo-2,4-dinitrobenzene
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])I

Specifications

Melting Point 87°C to 90°C
Quantity 5 g
Beilstein 1113584
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 294.01
Percent Purity 98%
Chemical Name or Material 1-Iodo-2,4-dinitrobenzene
compliance-icons
Product Identifier
  • 1-Iodo-2,4-dinitrobenzene
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 1-Iodo-2,4-dinitrobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.