missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-2,4-dimethoxybenzene, 97%

Product Code. p-7047548
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11328726

Brand: Thermo Scientific Alfa Aesar L19420.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20469-63-0 | C8H9IO2 | 264.062 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20469-63-0
Molecular Formula C8H9IO2
Molecular Weight (g/mol) 264.062
MDL Number MFCD00059268
InChI Key ZDUYJCKEORTAQE-UHFFFAOYSA-N
Synonym 2,4-dimethoxyiodobenzene, 2,4-dimethoxy-1-iodobenzene, 1,3-dimethoxy-4-iodobenzene, 1-iodo-2,4-dimethoxy-benzene, benzene, 1-iodo-2,4-dimethoxy, 4-iodo-1,3-dimethoxybenzene, pubchem3053, 4-iodo-3-methoxy anisole, dimethoxyiodobenzene, cambridge id 5107587
PubChem CID 140694
IUPAC Name 1-iodo-2,4-dimethoxybenzene
SMILES COC1=CC(=C(C=C1)I)OC

Specifications

Melting Point 37°C to 41°C
Boiling Point 135°C (14 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.61
Quantity 1 g
Beilstein 1945135
Sensitivity Light sensitive
Formula Weight 264.06
Percent Purity 97%
Chemical Name or Material 1-Iodo-2,4-dimethoxybenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.