missing translation for 'onlineSavingsMsg'
Learn More

1-Indoleacetic acid, 95%

Product Code. 15440919
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15440919 250 mg 250mg
15420919 1 g 1g
15430919 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15440919 Supplier Thermo Scientific Alfa Aesar Supplier No. H63935.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 24297-59-4 | C10H9NO2 | 175.187 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24297-59-4
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
MDL Number MFCD00047262
InChI Key WQJFIWXYPKYBTO-UHFFFAOYSA-N
Synonym 1h-indole-1-acetic acid, 2-1h-indol-1-yl acetic acid, indol-1-yl-acetic acid, indole-1-acetic acid, 1h-indol-1-ylacetic acid, indol-1-ylacetic acid, indole-n-acetic acid, 1-indoleacetic acid, pubchem20663, 1-indolyl acetic acid
PubChem CID 90448
ChEBI CHEBI:72814
IUPAC Name 2-indol-1-ylacetic acid
SMILES C1=CC=C2C(=C1)C=CN2CC(=O)O

Specifications

Quantity 250 mg
Beilstein 142924
Formula Weight 175.18
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 1-Indoleacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.