missing translation for 'onlineSavingsMsg'
Learn More

1-Indanone, 99+%

Product Code. 11924851 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Unit Size:
100g
10g
25g
500g
This item is not returnable. View return policy

Product Code. 11924851

Brand: Thermo Scientific Acros 122115000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 83-33-0 | C9H8O | 132.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-33-0
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.16
InChI Key QNXSIUBBGPHDDE-UHFFFAOYSA-N
Synonym 1-indanone, indan-1-one, 2,3-dihydro-1h-inden-1-one, indanone, alpha-indanone, alpha-hydrindone, 1h-inden-1-one, 2,3-dihydro, 1-indone, hydrindone, .alpha.-indanone
PubChem CID 6735
ChEBI CHEBI:17404
IUPAC Name 2,3-dihydroinden-1-one
SMILES C1CC(=O)C2=CC=CC=C21

Specifications

Melting Point 38.0°C to 42.0°C
Color Yellow to Brown
Density 1.1000g/mL
Boiling Point 243.0°C to 245.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass Bottle
Quantity 500 g
Beilstein 07,36
Specific Gravity 1.1
Solubility Information Solubility in water: 6.5g/L (20°C). Other solubilities: soluble in ethanol,ether and chloroform
Formula Weight 132.16
Percent Purity 99+%
Physical Form Crystalline Mass
Chemical Name or Material 1-Indanone
Show More Show Less
compliance-icons
Product Identifier
  • 1-Indanone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.