Learn More
1-Indanone, 99+%, Thermo Scientific Chemicals
£20.80 - £710.00
Chemical Identifiers
CAS | 83-33-0 |
---|---|
Molecular Formula | C9H8O |
Molecular Weight (g/mol) | 132.162 |
MDL Number | MFCD00003785 |
InChI Key | QNXSIUBBGPHDDE-UHFFFAOYSA-N |
Synonym | 1-indanone, indan-1-one, 2,3-dihydro-1h-inden-1-one, indanone, alpha-indanone, alpha-hydrindone, 1h-inden-1-one, 2,3-dihydro, 1-indone, hydrindone, .alpha.-indanone |
PubChem CID | 6735 |
ChEBI | CHEBI:17404 |
IUPAC Name | 2,3-dihydroinden-1-one |
SMILES | C1CC(=O)C2=CC=CC=C21 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11414170
|
Thermo Scientific Alfa Aesar
A11056.06 |
5 g |
£20.80
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11438903
|
Thermo Scientific Alfa Aesar
A11056.14 |
25 g |
£46.90
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11448903
|
Thermo Scientific Alfa Aesar
A11056.22 |
100 g |
£141.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11458903
|
Thermo Scientific Alfa Aesar
A11056.36 |
500 g |
£710.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Indanone is an oxidation product of indan, a component of fuels, solvents and varnishes. 1-Indanone is also a metabolite of Thalidomide that has been shown to inhibit the attachment of tumor cells to concanavalin A coated plastic surfaces.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Indanone is an oxidation product of indan, a component of fuels, solvents and varnishes. 1-Indanone is also a metabolite of Thalidomide that has been shown to inhibit the attachment of tumor cells to concanavalin A coated plastic surfaces.
Solubility
Soluble in water (6.5 g/L at 20°C).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
83-33-0 | |
132.162 | |
QNXSIUBBGPHDDE-UHFFFAOYSA-N | |
6735 | |
2,3-dihydroinden-1-one |
C9H8O | |
MFCD00003785 | |
1-indanone, indan-1-one, 2,3-dihydro-1h-inden-1-one, indanone, alpha-indanone, alpha-hydrindone, 1h-inden-1-one, 2,3-dihydro, 1-indone, hydrindone, .alpha.-indanone | |
CHEBI:17404 | |
C1CC(=O)C2=CC=CC=C21 |
Specifications
83-33-0 | |
1.103 | |
113°C (235°F) | |
MFCD00003785 | |
1-indanone, indan-1-one, 2,3-dihydro-1h-inden-1-one, indanone, alpha-indanone, alpha-hydrindone, 1h-inden-1-one, 2,3-dihydro, 1-indone, hydrindone, .alpha.-indanone | |
QNXSIUBBGPHDDE-UHFFFAOYSA-N | |
2,3-dihydroinden-1-one | |
6735 | |
132.16 | |
1-Indanone |
38°C to 42°C | |
243°C to 245°C | |
C9H8O | |
507957 | |
Soluble in water (6.5g/L at 20°C). | |
C1CC(=O)C2=CC=CC=C21 | |
132.162 | |
CHEBI:17404 | |
≥99% |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
missing translation for 'einecsNumber' : 201-470-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only