missing translation for 'onlineSavingsMsg'
Learn More

1-Indanone, 99+%

Product Code. 11448903
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448903 100 g 100g
11414170 5 g 5g
11438903 25 g 25g
11458903 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11448903 Supplier Thermo Scientific Alfa Aesar Supplier No. A11056.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 83-33-0 | C9H8O | 132.162 g/mol

1-Indanone is an oxidation product of indan, a component of fuels, solvents and varnishes. 1-Indanone is also a metabolite of Thalidomide that has been shown to inhibit the attachment of tumor cells to concanavalin A coated plastic surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Indanone is an oxidation product of indan, a component of fuels, solvents and varnishes. 1-Indanone is also a metabolite of Thalidomide that has been shown to inhibit the attachment of tumor cells to concanavalin A coated plastic surfaces.

Solubility
Soluble in water (6.5 g/L at 20°C).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-33-0
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.162
MDL Number MFCD00003785
InChI Key QNXSIUBBGPHDDE-UHFFFAOYSA-N
Synonym 1-indanone, indan-1-one, 2,3-dihydro-1h-inden-1-one, indanone, alpha-indanone, alpha-hydrindone, 1h-inden-1-one, 2,3-dihydro, 1-indone, hydrindone, .alpha.-indanone
PubChem CID 6735
ChEBI CHEBI:17404
IUPAC Name 2,3-dihydroinden-1-one
SMILES C1CC(=O)C2=CC=CC=C21

Specifications

Melting Point 38°C to 42°C
Density 1.103
Boiling Point 243°C to 245°C
Flash Point 113°C (235°F)
Quantity 100 g
Beilstein 507957
Solubility Information Soluble in water (6.5g/L at 20°C).
Formula Weight 132.16
Percent Purity ≥99%
Chemical Name or Material 1-Indanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.