missing translation for 'onlineSavingsMsg'
Learn More

1-Hydroxypyrene Beta-D-Glucuronide, TRC

Product Code. 30010477
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010477 5 mg 5mg
30009731 1 mg 1mg
30008924 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010477 Supplier Toronto Research Chemicals Supplier No. TRCH9527505MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154717-05-2
Molecular Formula C22 H18 O7
Molecular Weight (g/mol) 394.37
Synonym 1-Hydroxy-pyrene-glucuronide
IUPAC Name (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-pyren-1-yloxyoxane-2-carboxylic acid
SMILES O[C@H]1[C@H](Oc2ccc3ccc4cccc5ccc2c3c45)O[C@@H]([C@@H](O)[C@@H]1O)C(=O)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C22H18O7/c23-17-18(24)20(21(26)27)29-22(19(17)25)28-14-9-7-12-5-4-10-2-1-3-11-6-8-13(14)16(12)15(10)11/h1-9,17-20,22-25H,(H,26,27)/t17-,18-,19+,20-,22+/m0/s1
Formula Weight 394.1053
Percent Purity >95%
Chemical Name or Material 1-Hydroxypyrene beta-D-Glucuronide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.