missing translation for 'onlineSavingsMsg'
Learn More

1-Hydroxypyrene beta-D-Glucuronide Methyl Ester, TRC

Product Code. 30401806
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401806 1 mg 1mg
30407565 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30401806 Supplier Toronto Research Chemicals Supplier No. TRCH9527601MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Hydroxypyrene beta-D-Glucuronide Methyl Ester
CAS 27735-78-0
Molecular Formula C23 H20 O7
Quantity 1 mg
Synonym 1-Pyrenyl beta-D-Glucopyranosiduronic Acid Methyl Ester
InChI Formula InChI=1S/C23H20O7/c1-28-22(27)21-19(25)18(24)20(26)23(30-21)29-15-10-8-13-6-5-11-3-2-4-12-7-9-14(15)17(13)16(11)12/h2-10,18-21,23-26H,1H3
SMILES COC(=O)C1OC(Oc2ccc3ccc4cccc5ccc2c3c45)C(O)C(O)C1O
IUPAC Name methyl 3,4,5-trihydroxy-6-pyren-1-yloxy-tetrahydropyran-2-carboxylate
Molecular Weight (g/mol) 408.4
Formula Weight 408.1209
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.