missing translation for 'onlineSavingsMsg'
Learn More

1-Hydroxypyrene Beta-D-Glucuronide (1.0 mg/mL in Methanol), TRC

Product Code. 30478931
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30478931 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30478931 Supplier Toronto Research Chemicals Supplier No. TRCKIT98751X1ML
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 154717-05-2
Molecular Formula C22 H18 O7
Quantity 1 mL
Synonym 1-Hydroxy-pyrene-glucuronide
InChI Formula InChI=1S/C22H18O7/c23-17-18(24)20(21(26)27)29-22(19(17)25)28-14-9-7-12-5-4-10-2-1-3-11-6-8-13(14)16(12)15(10)11/h1-9,17-20,22-25H,(H,26,27)/t17-,18-,19+,20-,22+/m0/s1
SMILES O[C@H]1[C@H](Oc2ccc3ccc4cccc5ccc2c3c45)O[C@@H]([C@@H](O)[C@@H]1O)C(=O)O
IUPAC Name (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-pyren-1-yloxyoxane-2-carboxylic acid
Molecular Weight (g/mol) 394.37
Formula Weight 394.1053
Concentration or Composition (by Analyte or Components) 1-Hydroxypyrene beta-D-Glucuronide 1 mg/mL
Chemical Name or Material 1-Hydroxypyrene beta-D-Glucuronide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.