missing translation for 'onlineSavingsMsg'
Learn More

1-Hydroxyethylidenebis(phosphonic acid), 96%

Product Code. 11470590
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470590 50 g 50g
11315847 2 g 2g
11305847 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11470590 Supplier Thermo Scientific Alfa Aesar Supplier No. 043502.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2809-21-4 | C2H8O7P2 | 206.027 g/mol

1-Hydroxyethylidenebis(phosphonic acid) acts as human protein tyrosine phosphatase inhibitor. It is also useful for photographic applications. It finds application to strengthen bone, osteoporosis treatment and to treat Paget's disease of bone. In addition, it is used in corrosion inhibition in circulating cool water systems, oil fields and low-pressure boilers with electric power, the chemical industry, metallurgy and fertilizers. It is also utilized for formulating reverse osmosis and thermal desalination anti-scaling agents. Furthermore, it is used in detergents, water treatment, cosmetics and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2809-21-4
Molecular Formula C2H8O7P2
Molecular Weight (g/mol) 206.027
MDL Number MFCD00070585
InChI Key DBVJJBKOTRCVKF-UHFFFAOYSA-N
Synonym etidronic acid, etidronate, hedp, 1-hydroxyethylidene-1,1-diphosphonic acid, ehdp, etidronsaeure, acetodiphosphonic acid, hydroxyethanediphosphonic acid, oxyethylidenediphosphonic acid, turpinal sl
PubChem CID 3305
ChEBI CHEBI:4907
IUPAC Name (1-hydroxy-1-phosphonoethyl)phosphonic acid
SMILES CC(O)(P(=O)(O)O)P(=O)(O)O

Specifications

Odor Odorless
Quantity 50 g
UN Number UN3261
Merck Index 14,3863
Solubility Information Soluble in water.
Formula Weight 206.03
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 1-Hydroxyethylidenebis (phosphonic acid)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.