missing translation for 'onlineSavingsMsg'
Learn More

1-Hydroxyethylethoxypiperazine, 95%

Product Code. 10249343 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10249343 100 mL Glass bottle 100mL
10287490 5 mL Glass bottle 5mL
10023250 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10249343 Supplier Thermo Scientific Acros Supplier No. 259111000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13349-82-1 | C8H18N2O2 | 174.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13349-82-1
Molecular Formula C8H18N2O2
Molecular Weight (g/mol) 174.24
MDL Number MFCD00074931
InChI Key FLNQAPQQAZVRDA-UHFFFAOYSA-N
Synonym 1-2-2-hydroxyethoxy ethyl piperazine, 2-2-piperazin-1-yl ethoxy ethanol, 2-2-piperazin-1-ylethoxy ethanol, 2-2-1-piperazinyl ethoxy ethanol, 1-2-2-hydroxyethoxyl-ethyl piperazine, 1-2-2-hydroxyethoxy-ethyl-piperazine, ethanol, 2-2-1-piperazinyl ethoxy
PubChem CID 139436
IUPAC Name 2-(2-piperazin-1-ylethoxy)ethanol
SMILES C1CN(CCN1)CCOCCO

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Brown-Yellow to Colorless
Density 1.0100g/mL
Boiling Point 150.0°C (13.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4960 to 1.4980
Quantity 100 mL
Specific Gravity 1.01
Formula Weight 174.24
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1-Hydroxyethylethoxypiperazine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.