missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Hydroxy-4-(p-tolylamino)anthracene-9,10-dione, TRC
Specifications
Specifications
| Chemical Name or Material | Solvent Violet 13 |
| CAS | 81-48-1 |
| Molecular Formula | C21 H15 N O3 |
| Quantity | 1 g |
| Synonym | 1-Hydroxy-4-(4-methylanilino)anthraquinone, 1-Hydroxy-4-(p-toluidino)anthraquinone, 1-Hydroxy-4-(p-tolylamino)-9,10-anthraquinone, 1-Hydroxy-4-[(4-methylphenyl)-9,10-anthracenedione, 1-Hydroxy-4-p-toluidino-9,10-anthraquinone, 1-p-Toluidino-4-hydroxyanthraquinone, 11092 Violet, Ahcoquinone Blue IR Base, Alizarin violet 3B, Alizarine Irisol R Base, Alizarine Purple Lake SS, Alizarine Violet 3B, Alizarine Violet 3B Base, Alizurol Purple, Alizurol Purple SS, C.I. 60725, C.I. Disperse Blue 72, C.I. Solvent Blue 90, C.I. Solvent Violet 13, Calco Oil Violet ZIRS, D and C Violet No. 2, D&C Violet 2, D&C Violet No. 2, Diaresin Blue G, Disperse Blue 72, Dispersol Violet B-G, Duranol Brilliant Violet TG, Irisol base, Japan Purple 201, Japan Violet 201, Japan Violet No. 201, KP Plast Violet 2R, Kayaset Blue A 2R, Lurafix Blue 590, Macrolex Violet 2R, Macrolex Violet B, N-(4-Hydroxy-1-anthraquinonyl)-4-methylaniline, NSC 2856, Oil Violet IRS, Oil Violet ZIRS, Oplas Violet 730, Orient Oil Violet 730, Plast Violet 8840, Purple No. 201, Pyla-Cert D and C Violet 2, Quinizarin Blue, Resiren Blue TR, Resolin Blue RRL, Sandoplast Violet RSB, Solvaperm Blue B, Solvent Blue 90, Solvent Violet 13, Sumikaron Violet 3BL, Sumikaron Violet B, Sumiplast Violet B, Teraprint Blue 6R, Thermoplast Blue 684, Transetile Violet P 3B, Violet 2, Violet No. 201, Waxoline Purple A |
| InChI Formula | InChI=1S/C21H15NO3/c1-12-6-8-13(9-7-12)22-16-10-11-17(23)19-18(16)20(24)14-4-2-3-5-15(14)21(19)25/h2-11,22-23H,1H3 |
| SMILES | Cc1ccc(Nc2ccc(O)c3C(=O)c4ccccc4C(=O)c23)cc1 |
| IUPAC Name | 1-hydroxy-4-(4-methylanilino)anthracene-9,10-dione |
| Molecular Weight (g/mol) | 329.35 |
| Formula Weight | 329.1052 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?