missing translation for 'onlineSavingsMsg'
Learn More

1-Hydroxy-3-(oleoyloxy)propan-2-yl (9Z,12Z,15Z)-octadeca-9,12,15-trienoate, TRC

Product Code. 30399209
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399209 1 mg 1mg
30400836 10 mg -
30405141 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30399209 Supplier Toronto Research Chemicals Supplier No. TRCH3244301MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149249-80-9
Molecular Formula C39H68O5
Molecular Weight (g/mol) 616.95
Synonym 1-(Hydroxymethyl)-2-[[(9Z)-1-oxo-9-octadecen-1-yl]oxy]ethyl (9Z,12Z,15Z)-9,12,15-octadecatrienoate, (all-Z)-(±)-1-(Hydroxymethyl)-2-[(1-oxo-9-octadecenyl)oxy]ethyl 9,12,15-Octadecatrienoic Acid
IUPAC Name [3-hydroxy-2-[(9Z,12Z,15Z)-octadeca-9,12,15-trienoyl]oxypropyl] (Z)-octadec-9-enoate
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCC(CO)OC(=O)CCCCCCC\C=C/C\C=C/C\C=C/CC

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C39H68O5/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(41)43-36-37(35-40)44-39(42)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h6,8,12,14,17-20,37,40H,3-5,7,9-11,13,15-16,21-36H2,1-2H3/b8-6-,14-12-,19-17-,20-18-
Formula Weight 616.5067
Chemical Name or Material 1-Hydroxy-3-(oleoyloxy)propan-2-yl (9Z,12Z,15Z)-octadeca-9,12,15-trienoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.