missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[1-Hydroxy-3-(methylisopentylamino)propylidene] Bisphosphonic Acid Monosodium Salt(Ibandronic Acid Impurity), TRC
Specifications
Specifications
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C9H23NO7P2.Na/c1-8(2)4-6-10(3)7-5-9(11,18(12,13)14)19(15,16)17;/h8,11H,4-7H2,1-3H3,(H2,12,13,14)(H2,15,16,17);/q;+1/p-1 |
| Chemical Name or Material | [1-Hydroxy-3-(methylisopentylamino)propylidene] Bisphosphonic Acid Monosodium Salt (Ibandronic Acid Impurity) |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?