missing translation for 'onlineSavingsMsg'
Learn More

[1-Hydroxy-3-(methylisopentylamino)propylidene] Bisphosphonic Acid Monosodium Salt(Ibandronic Acid Impurity), TRC

Product Code. 30394210
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394210 10 mg 10mg
30399779 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30394210 Supplier Toronto Research Chemicals Supplier No. TRCH94695510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1797984-36-1
Molecular Formula C9H22NNaO7P2
Molecular Weight (g/mol) 341.21
Synonym [1-Hydroxy-3-(Methylisopentylamino)Propylidene] Bisphosphonic Acid Monosodium Salt, (Ibandronic Acid Impurity)
SMILES CCCCCCCCCCCCCCCCCCCC(O)=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C9H23NO7P2.Na/c1-8(2)4-6-10(3)7-5-9(11,18(12,13)14)19(15,16)17;/h8,11H,4-7H2,1-3H3,(H2,12,13,14)(H2,15,16,17);/q;+1/p-1
Chemical Name or Material [1-Hydroxy-3-(methylisopentylamino)propylidene] Bisphosphonic Acid Monosodium Salt (Ibandronic Acid Impurity)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.