missing translation for 'onlineSavingsMsg'
Learn More

1-Hydroxy-2-naphthoic acid, 98%

Product Code. 10346550 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10346550 100 g Plastic bottle 100g
10183000 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10346550 Supplier Thermo Scientific Acros Supplier No. 149231000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 86-48-6 | C11H8O3 | 188.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-48-6
Molecular Formula C11H8O3
Molecular Weight (g/mol) 188.18
MDL Number MFCD00003960
InChI Key SJJCQDRGABAVBB-UHFFFAOYSA-N
Synonym 1-hydroxy-2-naphthoic acid, 1-naphthol-2-carboxylic acid, 2-carboxy-1-naphthol, 2-naphthalenecarboxylic acid, 1-hydroxy, 1-hydroxy-2-naphthoate, 1-hydroxy-2-naphthalenecarboxylic acid, unii-u8lz3r07l8, 2-naphthoic acid, 1-hydroxy, alpha-hydroxynaphthoic acid, 1-hydroxy-naphthalene-2-carboxylic acid
PubChem CID 6844
ChEBI CHEBI:36108
IUPAC Name 1-hydroxynaphthalene-2-carboxylic acid
SMILES C1=CC=C2C(=C1)C=CC(=C2O)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 199.0°C to 205.0°C
Color Tan to White
Flash Point >150°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 100 g
Beilstein 10, 331
Merck Index 15, 4873
Solubility Information Solubility in water: soluble in hot water. Other solubilities: practically insoluble in cold water, soluble in benzene and chloroform, freely soluble in alcohol and ether
Formula Weight 188.18
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material 1-Hydroxy-2-naphthoic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Hydroxy-2-naphthoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.