missing translation for 'onlineSavingsMsg'
Learn More

1-Hydroxy-2,7-diamino Mitosene (Mixture cis/trans), TRC

Product Code. 30132634
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
10 mg
Unit Size:
10mg
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132634 2.5 mg 2.5mg
30004190 1 mg 1mg
30004335 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132634 Supplier Toronto Research Chemicals Supplier No. TRCH9397002.5MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1192552-64-9
Molecular Formula C14 H16 N4 O5
Molecular Weight (g/mol) 320.3
Synonym 1H-Pyrrolo[1,2-a]indole-5,8-dione, 2,7-diamino-9-[[(aminocarbonyl)oxy]methyl]-2,3-dihydro-1-hydroxy-6-methyl-, (2S)-, 1-Hydroxy-2,7-diamino Mitosene (Mixture cis/trans), (1RS,2S)-1-Hydroxy-2,7-diaminomitosene, (2S)-2,7-Diamino-9-[[(aminocarbonyl)oxy]methyl]-2,3-dihydro-1-hydroxy-6-methyl-1H-pyrrolo[1,2-a]indole-5,8-dione
IUPAC Name [(2S)-2,6-diamino-3-hydroxy-7-methyl-5,8-dioxo-2,3-dihydro-1H-pyrrolo[1,2-a]indol-4-yl]methyl carbamate
SMILES CC1=C(N)C(=O)c2c(COC(=O)N)c3C(O)[C@@H](N)Cn3c2C1=O

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C14H16N4O5/c1-4-8(16)13(21)7-5(3-23-14(17)22)9-12(20)6(15)2-18(9)10(7)11(4)19/h6,12,20H,2-3,15-16H2,1H3,(H2,17,22)/t6-,12?/m0/s1
Formula Weight 320.1121
Chemical Name or Material (1RS,2S)-1-Hydroxy-2,7-diaminomitosene (cis,trans mixture)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.