missing translation for 'onlineSavingsMsg'
Learn More

1-Hydroxy-1-cyclopropanecarboxylic acid, 97%

Product Code. 10733151 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10733151 1 g Glass bottle 1g
11905081 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10733151 Supplier Thermo Scientific Acros Supplier No. 301430010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17994-25-1 | C4H5O3 | 101.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17994-25-1
Molecular Formula C4H5O3
Molecular Weight (g/mol) 101.08
MDL Number MFCD00010797
InChI Key GQXURJDNDYACGE-UHFFFAOYSA-M
Synonym 1-hydroxy-1-cyclopropanecarboxylic acid, 1-hydroxycyclopropanecarboxylic acid, 1-hydroxy-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-hydroxy, 1-hydroxy-1-cyclopropyl carboxylic acid, pubchem12465, acmc-20a5ho, 1-carboxy-1-hydroxycyclopropane, 1-hydroxycyclopropanecaboxylic acid, 1-hydroxycyclo-propanecarboxylic acid
PubChem CID 2733160
IUPAC Name 1-hydroxycyclopropane-1-carboxylic acid
SMILES OC1(CC1)C([O-])=O

Specifications

Melting Point 108.0°C to 110.0°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Formula Weight 102.09
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material 1-Hydroxy-1-cyclopropanecarboxylic acid

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.