missing translation for 'onlineSavingsMsg'
Learn More

1-Hydroperoxy-1-phenylethane, TRC

Product Code. 30407411
Change view
Click to view available options
Quantity:
25 mg
500 mg
Unit Size:
25mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30407411 500 mg 500mg
30398879 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30407411 Supplier Toronto Research Chemicals Supplier No. TRCH950265500MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Hydroperoxy-1-phenylethane
CAS 3071-32-7
Molecular Formula C8H10O2
Quantity 500 mg
Synonym 1-Phenylethyl Hydroperoxide, (1-Hydroperoxyethyl)benzene, 1-Hydroperoxy-1-phenylethane, Ethylbenzene Hydroperoxide, Ethylbenzene α-Hydroperoxide, α-Hydroperoxyethylbenzene, α-Methylbenzyl Hydroperoxide, α-Phenylethyl Hydroperoxide
InChI Formula InChI=1S/C8H10O2/c1-7(10-9)8-5-3-2-4-6-8/h2-7,9H,1H3
SMILES CC(OO)C1=CC=CC=C1
IUPAC Name 1-hydroperoxyethylbenzene
Molecular Weight (g/mol) 138.16
Formula Weight 138.0681
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.