missing translation for 'onlineSavingsMsg'
Learn More

1-Hydrazinophthalazine hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 11426166
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11426166 25 g 25g
11416166 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11426166 Supplier Thermo Scientific Alfa Aesar Supplier No. B22995.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 304-20-1 | C8H9ClN4 | 196.638 g/mol

A monoamine oxidase inhibitor1-Hydrazinophthalazine hydrochloride is a non-nucleoside analog that inhibits DNA methylation and reactivates the expression of tumor suppressor genes. It acts as a non-selective MAO-A/B inhibitor and semicarbazide sensitive amine oxidase inhibitor. Further, it serves as an antihypertensive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A monoamine oxidase inhibitor1-Hydrazinophthalazine hydrochloride is a non-nucleoside analog that inhibits DNA methylation and reactivates the expression of tumor suppressor genes. It acts as a non-selective MAO-A/B inhibitor and semicarbazide sensitive amine oxidase inhibitor. Further, it serves as an antihypertensive.

Solubility
Soluble in water. Slightly soluble in diethyl ether and alcohol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 304-20-1
Molecular Formula C8H9ClN4
Molecular Weight (g/mol) 196.638
MDL Number MFCD00135998
InChI Key ZUXNZUWOTSUBMN-UHFFFAOYSA-N
Synonym hydralazine hydrochloride, 1-hydrazinophthalazine hydrochloride, 1-hydrazinylphthalazine hydrochloride, apresoline, aiselazine, hydralazine hcl, hydralazine chloride, hyperazin, hyperex, lopress
PubChem CID 9351
ChEBI CHEBI:31672
IUPAC Name phthalazin-1-ylhydrazine;hydrochloride
SMILES C1=CC=C2C(=C1)C=NN=C2NN.Cl

Specifications

Melting Point ∼275°C (decomposition)
Quantity 25 g
UN Number UN2811
Beilstein 3568329
Merck Index 14,4763
Formula Weight 196.64
Percent Purity 98%
Chemical Name or Material 1-Hydrazinophthalazine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.