missing translation for 'onlineSavingsMsg'
Learn More

1-Hexyne, TRC

Product Code. 30398705
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
Product Code. Quantity unitSize
30398705 5 mL 5mL
1 options
This item is not returnable. View return policy

Product Code. 30398705

Brand: Toronto Research Chemicals TRCH2814155ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Hexyne
CAS 693-02-7
Molecular Formula C6H10
Quantity 5 mL
Synonym 1-Hexyn, Butylacetylene, NSC 9709, n-Butylacetylene
InChI Formula InChI=1S/C6H10/c1-3-5-6-4-2/h1H,4-6H2,2H3
SMILES CCCCC#C
IUPAC Name hex-1-yne
Molecular Weight (g/mol) 82.14
Formula Weight 82.0783
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.