missing translation for 'onlineSavingsMsg'
Learn More

1-Hexylmagnesium chloride, 1M in MeTHF, Thermo Scientific™

Product Code. 15428057
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15428057 500 mL 500mL
15418057 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15428057 Supplier Thermo Scientific Alfa Aesar Supplier No. H51167.AP
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 44767-62-6
Molecular Formula C6H13ClMg
Molecular Weight (g/mol) 144.93
MDL Number MFCD06798087
InChI Key ODOOOEXDBNGOOF-UHFFFAOYSA-M
Synonym hexylmagnesium chloride, 1-hexylmagnesium chloride, hexylmagnesium chloride solution, hexyl magnesium chloride, n-hexylmagnesium chloride, chloro hexyl magnesium, odoooexdbngoof-uhfffaoysa-m, 1-hexylmagnesium chloride, 1m in methf, hexylmagnesium chloride2.0m solution, hexylmagnesium chloride solution, 2.0 m in thf
PubChem CID 10877247
SMILES CCCCCC[Mg]Cl

Specifications

Linear Formula CH3(CH2)4CH2MgCl
Quantity 500 mL
UN Number UN2924
Sensitivity Air and moisture sensitive
IUPAC Name magnesium;hexane;chloride
Formula Weight 144.93
Concentration or Composition (by Analyte or Components) 1M in MeTHF
Chemical Name or Material 1-Hexylmagnesium chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.