missing translation for 'onlineSavingsMsg'
Learn More

1-Hexene, TRC

Product Code. 30401696
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30401696 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30401696 Supplier Toronto Research Chemicals Supplier No. TRCH29485150G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Hexene
CAS 592-41-6
Molecular Formula C6 H12
Quantity 50 g
Synonym 1-Hexene, 1-n-Hexene, Dialen 6, Hexene, Linealene 6, NSC 74121, α-Hexene
InChI Formula InChI=1S/C6H12/c1-3-5-6-4-2/h3H,1,4-6H2,2H3
SMILES CCCCC=C
IUPAC Name hex-1-ene
Molecular Weight (g/mol) 84.16
Formula Weight 84.0939
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.