missing translation for 'onlineSavingsMsg'
Learn More

1-Hexanol, TRC

Product Code. 30401067
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401067 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30401067 Supplier Toronto Research Chemicals Supplier No. TRCH281220100ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Hexanol
CAS 111-27-3
Molecular Formula C6 H14 O
Quantity 100 mL
Synonym Hexan-1-ol, 1-Hexanol, Hexyl alcohol (8CI), 1-Hexyl alcohol, 1-Hydroxyhexane, Amylcarbinol, Caproyl alcohol, Epal 6, Hexanol, Kalcohl 0698, NSC 9254, Pentylcarbinol, n-Hexan-1-ol, n-Hexanol, n-Hexyl alcohol
InChI Formula InChI=1S/C6H14O/c1-2-3-4-5-6-7/h7H,2-6H2,1H3
SMILES CCCCCCO
IUPAC Name hexan-1-ol
Molecular Weight (g/mol) 102.17
Formula Weight 102.1045
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.