missing translation for 'onlineSavingsMsg'
Learn More

1-Hexanoic Acid, TRC

Product Code. 30390405
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390405 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30390405 Supplier Toronto Research Chemicals Supplier No. TRCH292145100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Caproic Acid
CAS 142-62-1
Molecular Formula C6 H12 O2
Quantity 100 g
Synonym Hexanoic acid (8CI, 9CI, ACI), 1-Hexanoic acid, 1-Pentanecarboxylic acid, Butylacetic acid, Caproic acid, Capronic acid, Hexoic acid, Hexylic acid, n-Caproic acid, n-Hexanoic acid, n-Hexoic acid, n-Hexylic acid, NAA 60, NSC 35598, NSC 8266, Pentylformic acid
InChI Formula InChI=1S/C6H12O2/c1-2-3-4-5-6(7)8/h2-5H2,1H3,(H,7,8)
SMILES CCCCCC(=O)O
IUPAC Name hexanoic acid
Molecular Weight (g/mol) 116.16
Formula Weight 116.0837
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.