missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Hexanoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | Caproic Acid |
| CAS | 142-62-1 |
| Molecular Formula | C6 H12 O2 |
| Quantity | 500 g |
| Synonym | Hexanoic acid (8CI, 9CI, ACI), 1-Hexanoic acid, 1-Pentanecarboxylic acid, Butylacetic acid, Caproic acid, Capronic acid, Hexoic acid, Hexylic acid, n-Caproic acid, n-Hexanoic acid, n-Hexoic acid, n-Hexylic acid, NAA 60, NSC 35598, NSC 8266, Pentylformic acid |
| InChI Formula | InChI=1S/C6H12O2/c1-2-3-4-5-6(7)8/h2-5H2,1H3,(H,7,8) |
| SMILES | CCCCCC(=O)O |
| IUPAC Name | hexanoic acid |
| Molecular Weight (g/mol) | 116.16 |
| Formula Weight | 116.0837 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?