missing translation for 'onlineSavingsMsg'
Learn More

1-Hexanethiol, 96%

Product Code. 10046103 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10046103 500 mL Glass bottle 500mL
10012930 5 mL Glass bottle 5mL
10676992 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10046103 Supplier Thermo Scientific Acros Supplier No. 215275000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-31-9 | C6H14S | 118.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-31-9
Molecular Formula C6H14S
Molecular Weight (g/mol) 118.24
MDL Number MFCD00004909
InChI Key PMBXCGGQNSVESQ-UHFFFAOYSA-N
Synonym 1-hexanethiol, hexyl mercaptan, n-hexyl mercaptan, hexanethiol, 1-hexylthiol, 1-mercaptohexane, hexylthiol, n-hexanethiol, n-hexylthiol, n-hexylmercaptan
PubChem CID 8106
IUPAC Name hexane-1-thiol
SMILES CCCCCCS

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point -81°C to 80°C
Color Colorless to Yellow
Density 0.8400g/mL
Boiling Point 150.0°C to 154.0°C
Flash Point 30°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)5SH
Refractive Index 1.4470 to 1.4510
Quantity 500 mL
Beilstein 01, 408
Specific Gravity 0.84
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohols, ethyl ether, hydrocarbons
Formula Weight 118.24
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 1-Hexanethiol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.