missing translation for 'onlineSavingsMsg'
Learn More

1-Hexanesulfonic Acid Sodium Salt, TRC

Product Code. 30136341
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136341 100 g 100g
30136297 10 g 10g
30006000 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136341 Supplier Toronto Research Chemicals Supplier No. TRCH294385100G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2832-45-3
Molecular Formula C6H13NaO3S
Molecular Weight (g/mol) 188.22
Synonym Sodium 1-hexanesulfonate
IUPAC Name sodium;hexane-1-sulfonate
SMILES [Na+].CCCCCCS(=O)(=O)[O-]

Specifications

Quantity 100 g
InChI Formula InChI=1S/C6H14O3S.Na/c1-2-3-4-5-6-10(7,8)9;/h2-6H2,1H3,(H,7,8,9);/q;+1/p-1
Formula Weight 188.0483
Chemical Name or Material 1-Hexanesulfonic acid sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.