missing translation for 'onlineSavingsMsg'
Learn More

1-hexanesulfonic acid, sodium salt hydrate, 99%, Thermo Scientific Chemicals

Catalog Number 10062740
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2832-45-3
C6H13NaO3S
188.22
MFCD00007542
QWSZRRAAFHGKCH-UHFFFAOYSA-M
sodium 1-hexanesulfonate, sodium hexane-1-sulfonate, sodium hexanesulfonate, 1-hexanesulfonic acid sodium salt, 1-hexanesulfonic acid, sodium salt, 1-hexanesulfonic acid, sodium salt 1:1, 1-hexane sulfonic acid sodium salt, hexyl sodium sulfonate, 1-hexanesulfonate, sodium, hexanesulfonic acid na-salt
23677630
sodium;hexane-1-sulfonate
[Na+].CCCCCCS([O-])(=O)=O
This item is not returnable. View return policy
2832-45-3
C6H13NaO3S
188.22
MFCD00007542
QWSZRRAAFHGKCH-UHFFFAOYSA-M
sodium 1-hexanesulfonate, sodium hexane-1-sulfonate, sodium hexanesulfonate, 1-hexanesulfonic acid sodium salt, 1-hexanesulfonic acid, sodium salt, 1-hexanesulfonic acid, sodium salt 1:1, 1-hexane sulfonic acid sodium salt, hexyl sodium sulfonate, 1-hexanesulfonate, sodium, hexanesulfonic acid na-salt
23677630
sodium;hexane-1-sulfonate
[Na+].CCCCCCS([O-])(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Absorbance 5.5 w/v% aq. soln., pH 3, H3PO4, 0.06 max. at 280nm, 0.1 max. at 220nm, 1 max. at 210nm, 5.5 w/v% aq. soln., pH 3, H3PO4 at 0nm
Melting Point >300.0°C
Color White
Density Negligible
Loss on Drying 9.6% max. (100°C, 3 hrs) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3(CH2)5SO3Na·xH2O
Quantity 5 g
Solubility Information (10% in water) Clear colorless
Formula Weight 188.22
Vapor Pressure Negligible
Percent Purity 99%
Physical Form Crystalline Flakes or Solid
Chemical Name or Material 1-Hexanesulfonic acid, sodium salt hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'