Learn More
1-Hexanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous, Thermo Scientific Chemicals
£157.00 - £438.00
Chemical Identifiers
CAS | 2832-45-3 |
---|---|
Molecular Formula | C6H13NaO3S |
Molecular Weight (g/mol) | 188.22 |
MDL Number | MFCD00007542 |
InChI Key | QWSZRRAAFHGKCH-UHFFFAOYSA-M |
Synonym | sodium 1-hexanesulfonate, sodium hexane-1-sulfonate, sodium hexanesulfonate, 1-hexanesulfonic acid sodium salt, 1-hexanesulfonic acid, sodium salt, 1-hexanesulfonic acid, sodium salt 1:1, 1-hexane sulfonic acid sodium salt, hexyl sodium sulfonate, 1-hexanesulfonate, sodium, hexanesulfonic acid na-salt |
PubChem CID | 23677630 |
IUPAC Name | sodium;hexane-1-sulfonate |
SMILES | [Na+].CCCCCCS([O-])(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10741294
|
Thermo Scientific Acros
396670250 |
25 g | Glass bottle |
£157.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10360094
|
Thermo Scientific Acros
396671000 |
100 g | Glass bottle |
£438.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ion pair chromatographyChemical Identifiers
2832-45-3 | |
188.22 | |
QWSZRRAAFHGKCH-UHFFFAOYSA-M | |
23677630 | |
[Na+].CCCCCCS([O-])(=O)=O |
C6H13NaO3S | |
MFCD00007542 | |
sodium 1-hexanesulfonate, sodium hexane-1-sulfonate, sodium hexanesulfonate, 1-hexanesulfonic acid sodium salt, 1-hexanesulfonic acid, sodium salt, 1-hexanesulfonic acid, sodium salt 1:1, 1-hexane sulfonic acid sodium salt, hexyl sodium sulfonate, 1-hexanesulfonate, sodium, hexanesulfonic acid na-salt | |
sodium;hexane-1-sulfonate |
Specifications
2832-45-3 | |
99.0 | |
>300°C | |
2% max. (150°C) | |
99+% | |
CH3(CH2)5SO3Na | |
sodium 1-hexanesulfonate, sodium hexane-1-sulfonate, sodium hexanesulfonate, 1-hexanesulfonic acid sodium salt, 1-hexanesulfonic acid, sodium salt, 1-hexanesulfonic acid, sodium salt 1:1, 1-hexane sulfonic acid sodium salt, hexyl sodium sulfonate, 1-hexanesulfonate, sodium, hexanesulfonic acid na-salt | |
QWSZRRAAFHGKCH-UHFFFAOYSA-M | |
sodium;hexane-1-sulfonate | |
23677630 | |
99+% | |
Crystalline Solid |
0.02 max. (C=0.5 M in water) at 260nm, 0.04 max. (C=0.5 M in water) at 230nm, 0.06 max. (C=0.5 M in water) at 220nm, 0.1 max. (C=0.5 M in water) at 210nm | |
100.0 | |
White | |
Authentic | |
C6H13NaO3S | |
MFCD00007542 | |
Solubility in water: 300g/l | |
[Na+].CCCCCCS([O-])(=O)=O | |
188.22 | |
188.22 | |
Ion Pair Chromatography | |
1-Hexanesulfonic acid, sodium salt, Anhydrous |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 220-601-3
- 1-Hexanesulfonic acid, sodium salt
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only