missing translation for 'onlineSavingsMsg'
Learn More

1-Hexane Sulfonic Acid, Sodium Salt, 98+%, Ion Pair Chromatography, for Electrochemical Determination, Fisher Chemical

Product Code. 10510151 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Plastic powder jar
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10510151 25 g Plastic powder jar 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10510151 Supplier Fisher Chemical Supplier No. H/0439/46
  • ONLINE EXCLUSIVE PRICE
    £236.00
    £204.00 / 25g
    Save £32.00
    13% Off
View all promotions
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 300°C
CAS 2832-45-3
Color White
Physical Form Powder/Solid
Quantity 25 g
Molecular Formula C6H13NaO3S
MDL Number MFCD00007542
Synonym sodium 1-hexanesulfonate, sodium hexane-1-sulfonate, sodium hexanesulfonate, 1-hexanesulfonic acid sodium salt, 1-hexanesulfonic acid, sodium salt, 1-hexanesulfonic acid, sodium salt 1:1, 1-hexane sulfonic acid sodium salt, hexyl sodium sulfonate, 1-hexanesulfonate, sodium, hexanesulfonic acid na-salt
InChI Key QWSZRRAAFHGKCH-UHFFFAOYSA-M
SMILES [Na+].CCCCCCS([O-])(=O)=O
IUPAC Name sodium;hexane-1-sulfonate
Molecular Weight (g/mol) 188.22
PubChem CID 23677630
Formula Weight 188.22g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons
Product Identifier
  • 1-Hexanesulfonic acid, sodium salt
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.