missing translation for 'onlineSavingsMsg'
Learn More

1-Hexane-d13-thiol, CDN

Product Code. 30323862
Change view
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30323862 0.1 g 0.1g
30318171 0.25 g 0.25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30323862 Supplier CDN Supplier No. CDND64190.1G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Hexanethiol-D13
CAS 1142922-50-6
Molecular Formula C6 D13 H S
Quantity 0.1 g
Synonym 1-Hexane-1,1,2,2,3,3,4,4,5,5,6,6,6-d13-thiol (ACI), 1-Hexane-d13-thiol, 1-Hexanethiol-d13, 1-Hexanethiol-D13, Hexyl-d13 Mercaptan
InChI Formula InChI=1 S/C6H14S/c1-2-3-4-5-6-7/h7H,2-6H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2
SMILES [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])S
IUPAC Name 1,1,2,2,3,3,4,4,5,5,6,6,6-tridecadeuteriohexane-1-thiol
Molecular Weight (g/mol) 131.3205
Formula Weight 131.1632 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.