missing translation for 'onlineSavingsMsg'
Learn More

1-Hexadecylamine, tech. 90%, remainder mainly 1-octadecylamine

Product Code. 11460627
Change view
Click to view available options
Quantity:
25 g
250 g
1000 g
Unit Size:
1000g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460627 250 g 250g
11448740 25 g 25g
11450627 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460627 Supplier Thermo Scientific Alfa Aesar Supplier No. B22459.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 143-27-1 | C16H35N | 241.46 g/mol

1-Hexadecylamine is used for producing resin, senior detergent, anti-caking agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Hexadecylamine is used for producing resin, senior detergent, anti-caking agent.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents, acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-27-1
Molecular Formula C16H35N
Molecular Weight (g/mol) 241.46
MDL Number MFCD00008158
InChI Key FJLUATLTXUNBOT-UHFFFAOYSA-N
Synonym hexadecylamine, 1-hexadecylamine, cetylamine, 1-hexadecanamine, 1-aminohexadecane, n-hexadecylamine, hexyldecylamine, palmitamine, palmitylamine, n-cetylamine
PubChem CID 8926
IUPAC Name hexadecan-1-amine
SMILES CCCCCCCCCCCCCCCCN

Specifications

Melting Point 42°C to 47°C
Density 0.813
Boiling Point 144°C (2 mmHg)
Flash Point 140°C (284°F)
Assay Percent Range 90%
Refractive Index 1.4496
Quantity 250 g
UN Number UN3259
Beilstein 1634065
Solubility Information Insoluble in water.
Formula Weight 241.46
Percent Purity 90%
Grade Technical
Assay remainder mainly 1-octadecylamine
Chemical Name or Material 1-Hexadecylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.