missing translation for 'onlineSavingsMsg'
Learn More

(1-Hexadecyl)triphenylphosphonium bromide, 98+%

Product Code. 11458740
Change view
Click to view available options
Quantity:
5 g
10 g
100 g
Unit Size:
100g
10g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11458740 5 g 5g
11432188 10 g 10g
11468740 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11458740 Supplier Thermo Scientific Alfa Aesar Supplier No. A15180.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14866-43-4 | C34H48BrP | 567.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Widely used as preservative, water treatment in agriculture. Also used as antistatic agent in textiles, fibers, leather, as flocculants, deodorant and as phase transfer catalyst.

Solubility
Soluble in ethanol. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14866-43-4
Molecular Formula C34H48BrP
Molecular Weight (g/mol) 567.64
MDL Number MFCD00051858
InChI Key UXMZNEHSMYESLH-UHFFFAOYSA-M
Synonym hexadecyltriphenylphosphonium bromide, 1-hexadecyl triphenylphosphonium bromide, cetyltriphenylphosphonium bromide, phosphonium, hexadecyltriphenyl-, bromide, hexadecyl triphenyl phosphanium bromide, hexadecyltriphenylphosphanium bromide, acmc-1boiz, hexadecyl triphenyl phosphonium bromide, hexadecyl triphenyl phosphonium chloride, 1-hexadecyl triphenylphosphoniumbromide
PubChem CID 3084210
IUPAC Name hexadecyl(triphenyl)phosphanium;bromide
SMILES [Br-].CCCCCCCCCCCCCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 99°C to 101°C
Quantity 5 g
Beilstein 3582592
Sensitivity Hygroscopic
Solubility Information Soluble in ethanol. Slightly soluble in water.
Formula Weight 567.64
Percent Purity ≥98%
Chemical Name or Material (1-Hexadecyl)triphenylphosphonium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.