missing translation for 'onlineSavingsMsg'
Learn More

(1-Hexadecyl)trimethylammonium chloride, 96%

Product Code. 11394827
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
This item is not returnable. View return policy

Product Code. 11394827

Brand: Thermo Scientific Alfa Aesar 041861.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-02-7 | C19H42ClN | 320.00 g/mol

(1-Hexadecyl)trimethylammonium chloride is used as a topical antiseptic and surfactant. It is used in hair care formulations, cream rinse ingredient, hair conditioners and shampoos as a conditioning agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-02-7
Molecular Formula C19H42ClN
Molecular Weight (g/mol) 320.00
MDL Number MFCD00011773
InChI Key WOWHHFRSBJGXCM-UHFFFAOYSA-M
Synonym hexadecyltrimethylammonium chloride, cetrimonium chloride, cetyltrimethylammonium chloride, n,n,n-trimethylhexadecan-1-aminium chloride, dehyquart a, cetyl trimethyl ammonium chloride, aliquat 6, trimethylcetylammonium chloride, arquad 16-50, n-hexadecyltrimethylammonium chloride
PubChem CID 8154
ChEBI CHEBI:53581
IUPAC Name hexadecyl(trimethyl)azanium;chloride
SMILES [Cl-].CCCCCCCCCCCCCCCC[N+](C)(C)C

Specifications

Melting Point 234°C
Color White
Density 0.968 g/mL
Odor Odorless
Linear Formula CH3(CH2)15N(CH3)3Cl
Quantity 100 g
UN Number UN1759
Beilstein 3657974
Solubility Information Soluble in water.
Formula Weight 320
Percent Purity 96%
Physical Form Powder
Chemical Name or Material (1-Hexadecyl)trimethylammonium chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.