missing translation for 'onlineSavingsMsg'
Learn More

(1-Hexadecyl)trimethylammonium bromide, 98%, Thermo Scientific Chemicals

£25.50 - £501.00

Chemical Identifiers

CAS 57-09-0
Molecular Formula C19H42BrN
Molecular Weight (g/mol) 364.46
MDL Number MFCD00011772
InChI Key LZZYPRNAOMGNLH-UHFFFAOYSA-M
Synonym cetrimonium bromide, cetyltrimethylammonium bromide, hexadecyltrimethylammonium bromide, ctab, centimide, cetab, bromat, ctmab, cee dee, cetavlon
PubChem CID 5974
ChEBI CHEBI:3567
SMILES [Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11478740
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A15235.22
100 g
£25.50
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11488740
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A15235.36
500 g
£74.20
500g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11482268
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A15235.0I
5000 g
£501.00
5000g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

(1-Hexadecyl)trimethylammonium bromide is a cationic surfactant in hair conditioning products. It acts as a phase-transfer catalyst for the preparation of polyamide, polycarbonate and polythiocarbonate. It facilitates ligand-free palladium salt-catalyzed Heck and Suzuki cross-coupling reactions in water. It is an active component of the topical antiseptic cetrimide and used in household products such as shampoos and cosmetics. As a buffer solution, it is involved in the extraction of DNA as well as in the synthesis of gold nanoparticles and silican nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1-Hexadecyl)trimethylammonium bromide is a cationic surfactant in hair conditioning products. It acts as a phase-transfer catalyst for the preparation of polyamide, polycarbonate and polythiocarbonate. It facilitates ligand-free palladium salt-catalyzed Heck and Suzuki cross-coupling reactions in water. It is an active component of the topical antiseptic cetrimide and used in household products such as shampoos and cosmetics. As a buffer solution, it is involved in the extraction of DNA as well as in the synthesis of gold nanoparticles and silican nanoparticles.

Solubility
Soluble in alcohol, chloroform and water. Slightly soluble in acetone. Insoluble in diethyl ether and benzene.

Notes
Hygroscopic. Store in an airtight container. Incompatible with bases and strong oxidizing agents.
Specifications

Chemical Identifiers

57-09-0
364.46
LZZYPRNAOMGNLH-UHFFFAOYSA-M
5974
[Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C
C19H42BrN
MFCD00011772
cetrimonium bromide, cetyltrimethylammonium bromide, hexadecyltrimethylammonium bromide, ctab, centimide, cetab, bromat, ctmab, cee dee, cetavlon
CHEBI:3567

Specifications

57-09-0
5 to 7
Odorless
MFCD00011772
3598189
14,2025
Soluble in alcohol,chloroform and water. Slightly soluble in acetone. Insoluble in diethyl ether and benzene.
[Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C
364.46
CHEBI:3567
98%
∼245°C (decomposition)
244°C (471°F)
C19H42BrN
UN3077
Hygroscopic
cetrimonium bromide, cetyltrimethylammonium bromide, hexadecyltrimethylammonium bromide, ctab, centimide, cetab, bromat, ctmab, cee dee, cetavlon
LZZYPRNAOMGNLH-UHFFFAOYSA-M
hexadecyl(trimethyl)azanium;bromide
5974
364.46
(1-Hexadecyl)trimethylammonium bromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P314-P330-P332+P313-P362-P501c

H302-H315-H318-H335-H373

missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

missing translation for 'einecsNumber' : 200-311-3

missing translation for 'rtecsNumber' : BQ7875000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • (1-Hexadecyl)trimethylammonium bromide

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H373-May cause damage to organs.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only