missing translation for 'onlineSavingsMsg'
Learn More

(1-Hexadecyl)pyridinium chloride monohydrate, 98%

Product Code. 11405603
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11405603

Brand: Thermo Scientific Alfa Aesar A13499.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6004-24-6 | C21H40ClNO | 358.01 g/mol

(1-Hexadecyl)pyridinium chloride monohydrate is used in a study to assess removal of emulsified oil from oily waste water and removal of emulsified food and mineral oils from waste water. And it is used for detergent-based extraction of membrane-associated enzymes such as alkaline phosphatase, and in extraction of chromium and palladium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1-Hexadecyl)pyridinium chloride monohydrate is used in a study to assess removal of emulsified oil from oily waste water and removal of emulsified food and mineral oils from waste water. And it is used for detergent-based extraction of membrane-associated enzymes such as alkaline phosphatase, and in extraction of chromium and palladium.

Solubility
Soluble in water, alcohol, chloroform

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6004-24-6
Molecular Formula C21H40ClNO
Molecular Weight (g/mol) 358.01
MDL Number MFCD00149977
InChI Key NFCRBQADEGXVDL-UHFFFAOYSA-M
Synonym cetylpyridinium chloride monohydrate, 1-hexadecylpyridin-1-ium chloride hydrate, 1-hexadecylpyridinium chloride monohydrate, hexadecylpyridinium chloride monohydrate, unii-d9om4sk49p, pyridinium, 1-hexadecyl-, chloride, monohydrate, d9om4sk49p, cetylpyridinii chloridum, sprol tn
PubChem CID 22324
ChEBI CHEBI:3566
SMILES O.[Cl-].CCCCCCCCCCCCCCCCN1C=C[CH+]C=C1

Specifications

Melting Point 83°C to 86°C
Quantity 500 g
UN Number UN2811
Beilstein 3578606
Merck Index 14,2032
Solubility Information Soluble in water, alcohol, chloroform
IUPAC Name 1-hexadecyl-1,4-dihydropyridin-4-ylium hydrate chloride
Formula Weight 358.02 (339.99 Anhydrous)
Percent Purity 98%
Chemical Name or Material (1-Hexadecyl)pyridinium chloride monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.