missing translation for 'onlineSavingsMsg'
Learn More

1-Hexadecyl-3-methylimidazolium chloride monohydrate, 98%

Product Code. 10048741 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10048741 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10048741 Supplier Thermo Scientific Acros Supplier No. 356650100
  • ONLINE EXCLUSIVE PRICE
    £122.00
    £118.00 / 10g
    Save £4.00
    3% Off
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 404001-62-3 | C20H41ClN2O | 361.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Ionic liquid
TRUSTED_SUSTAINABILITY

Specifications

CAS 404001-62-3
CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Molecular Formula C20H41ClN2O
MDL Number MFCD07773044
Quantity 10 g
Synonym 1-hexadecyl-3-methyl-1h-imidazol-3-ium chloride hydrate, 1-hexadecyl-3-methylimidazolium chloride monohydrate, 1-hexadecyl-3-methyl-1h-imidazolium chloride hydrate
InChI Key YCRGEJFZTMXLMF-UHFFFAOYSA-M
SMILES O.[Cl-].CCCCCCCCCCCCCCCCN1C=C[N+](C)=C1
IUPAC Name 1-hexadecyl-3-methylimidazol-3-ium;chloride;hydrate
Molecular Weight (g/mol) 361.01
PubChem CID 60160661
Formula Weight 361.01
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 1-Hexadecyl-3-methylimidazolium chloride monohydrate
Show More Show Less
compliance-icons
Product Identifier
  • 1-Hexadecyl-3-methylimidazolium chloride monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.