missing translation for 'onlineSavingsMsg'
Learn More

1-Hexadecanol, 98%

Product Code. 11419123
Change view
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419123 500 g 500g
11429123 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419123 Supplier Thermo Scientific Alfa Aesar Supplier No. A11180.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 36653-82-4 | C16H34O | 242.45 g/mol

1-Hexadecanol is used as cosmetic ingredient, emollient, foam stabilizer. It is also used as the raw material for the manufacture of surfactants. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Hexadecanol is used as cosmetic ingredient, emollient, foam stabilizer. It is also used as the raw material for the manufacture of surfactants. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Soluble in alcohol, chloroform, ether

Notes
Keep away from direct sunlight. Incompatible with strong oxidizing agents, strong acids. Store at 2-8°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36653-82-4
Molecular Formula C16H34O
Molecular Weight (g/mol) 242.45
MDL Number MFCD00004760
InChI Key BXWNKGSJHAJOGX-UHFFFAOYSA-N
Synonym 1-hexadecanol, cetyl alcohol, hexadecanol, cetanol, palmityl alcohol, hexadecyl alcohol, n-cetyl alcohol, cetaffine, cetylol, cetal
PubChem CID 2682
ChEBI CHEBI:16125
IUPAC Name hexadecan-1-ol
SMILES CCCCCCCCCCCCCCCCO

Specifications

Melting Point 47°C to 51°C
Density 0.817 g/mL
Boiling Point 190°C (15 mm)
Flash Point 135°C (275°F)
Assay Percent Range 98%
Quantity 500 g
Beilstein 1748475
Merck Index 14,2028
Solubility Information Soluble in alcohol, chloroform, ether
Formula Weight 242.45
Percent Purity 98%
Chemical Name or Material 1-Hexadecanol
Show More Show Less
Product Identifier
  • 1-Hexadecanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.