missing translation for 'onlineSavingsMsg'
Learn More

1-Hexadecanol, 96%

Product Code. 11994601 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Unit Size:
10kg
1kg
2.5kg
This item is not returnable. View return policy

Product Code. 11994601

Brand: Thermo Scientific Acros 120480100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 36653-82-4 | C16H34O | 242.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36653-82-4
Molecular Formula C16H34O
Molecular Weight (g/mol) 242.45
MDL Number MFCD00004760
InChI Key BXWNKGSJHAJOGX-UHFFFAOYSA-N
Synonym 1-hexadecanol, cetyl alcohol, hexadecanol, cetanol, palmityl alcohol, hexadecyl alcohol, n-cetyl alcohol, cetaffine, cetylol, cetal
PubChem CID 2682
ChEBI CHEBI:16125
IUPAC Name hexadecan-1-ol
SMILES CCCCCCCCCCCCCCCCO

Specifications

Melting Point 47°C to 50°C
Color White
Boiling Point 344°C
Flash Point >157°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Fibre drum
Linear Formula CH3(CH2)15OH
Quantity 10 kg
Beilstein 01,429
Merck Index 15,2027
Solubility Information Solubility in water: insoluble in water. Other solubilities: slightly soluble in ethanol,soluble in acetone,very soluble in ether,benzene,chloroform
Viscosity 8 mPa.s (60°C)
Formula Weight 242.44
Percent Purity 96%
Physical Form Powder, Flakes or Pellets
Chemical Name or Material 1-Hexadecanol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.