missing translation for 'onlineSavingsMsg'
Learn More

1-Hexadecanesulfonyl chloride, 97+%

Product Code. 11437007
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11437007 50 g 50g
11427007 10 g 10g
11447007 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11437007 Supplier Thermo Scientific Alfa Aesar Supplier No. A14396.18
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 38775-38-1 | C16H33ClO2S | 324.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38775-38-1
Molecular Formula C16H33ClO2S
Molecular Weight (g/mol) 324.95
MDL Number MFCD00007459
InChI Key YOSVFFVBSPQTTP-UHFFFAOYSA-N
Synonym 1-hexadecanesulfonyl chloride, hexadecylsulfonyl chloride, 1-hexadecanesulfonylchloride, hexadecane-1-sulphonyl chloride, acmc-20aouo, hexadecanesulfonylchloride, hexadecanesulfonyl chloride, 1-hexadecylsulfonyl chloride, cetyl sulphonic acid chloride, 1-hexadecanesulphonyl chloride
PubChem CID 96923
IUPAC Name hexadecane-1-sulfonyl chloride
SMILES CCCCCCCCCCCCCCCCS(Cl)(=O)=O

Specifications

Melting Point 56°C to 58°C
Quantity 50 g
UN Number UN3261
Beilstein 1789245
Sensitivity Moisture sensitive
Formula Weight 324.96
Percent Purity 98%
Chemical Name or Material 1-Hexadecanesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.