missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ 1-Hex-GlcNAlk
GREENER_CHOICE

Product Code. 18739506
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18739506 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18739506 Supplier Tocris Bioscience™ Supplier No. 7751/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Metabolic chemical reporter for studying glycosylation and glycoproteins

1-Hex-GlcNAlk is a metabolic chemical reporter (MCR) for studying glycoproteins and glycosylation; it comprises GlcNAlk functionalized at the anomeric position with hexanoic acid. 1-Hex-GlcNAlk exhibits time- and concentration-dependent specific enzymatic labeling of proteins in HeLa cells, with negligible background labeling detected in cell lysates.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2 S,3 R,4 R,5 S,6 R)-4,5-Dihydroxy-6-(hydroxymethyl)-3-(pent-4-ynamido)tetrahydro-2 H-pyran-2-yl hexanoate
Quantity 10 mg
Target GlcNAc Detection
Molecular Formula C17H27NO7
Purity 0.95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.